Anti-CD70 Rat Monoclonal Antibody [clone: 118510]

Supplier: R & D Systems
MAB783
220021-549EA 763.57 USD
220021-549
Anti-CD70 Rat Monoclonal Antibody [clone: 118510]
Antibodies
Type: Primary
Antigen: CD70
Clonality: Monoclonal
Clone: 118510
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR