Anti-NCAM1 Rat Monoclonal Antibody [clone: 809220]

Supplier: R & D Systems
MAB7820
102887-384EA 594.37 USD
102887-384
Anti-NCAM1 Rat Monoclonal Antibody [clone: 809220]
Antibodies
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 809220
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR