Anti-IL22 Mouse Monoclonal Antibody [clone: 142938]

Supplier: R & D Systems
MAB782
220021-546EA 772.25 USD
220021-546
Anti-IL22 Mouse Monoclonal Antibody [clone: 142938]
Antibodies
Type: Primary
Antigen: IL22
Clonality: Monoclonal
Clone: 142938
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR