Anti-CCL19 Mouse Monoclonal Antibody [clone: 866202]

Supplier: R & D Systems
MAB7800
102887-382EA 577.02 USD
102887-382
Anti-CCL19 Mouse Monoclonal Antibody [clone: 866202]
Antibodies
Type: Primary
Antigen: CCL19
Clonality: Monoclonal
Clone: 866202
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR