Anti-IL1RN Mouse Monoclonal Antibody [clone: 114801]

Supplier: R & D Systems
MAB780
220021-544EA 772.25 USD
220021-544
Anti-IL1RN Mouse Monoclonal Antibody [clone: 114801]
Antibodies
Type: Primary
Antigen: IL1RN
Clonality: Monoclonal
Clone: 114801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR