Anti-MS4A4A Mouse Monoclonal Antibody [clone: 818112]

Supplier: R & D Systems
MAB7797
102887-380EA 594.37 USD
102887-380
Anti-MS4A4A Mouse Monoclonal Antibody [clone: 818112]
Antibodies
Type: Primary
Antigen: MS4A4A
Clonality: Monoclonal
Clone: 818112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR