Anti-SNAPIN Mouse Monoclonal Antibody [clone: 809116]

Supplier: R & D Systems
MAB7795
102887-376EA 577.02 USD
102887-376
Anti-SNAPIN Mouse Monoclonal Antibody [clone: 809116]
Antibodies
Type: Primary
Antigen: SNAPIN
Clonality: Monoclonal
Clone: 809116
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR