Anti-PTGIS Mouse Monoclonal Antibody [clone: 852307]

Supplier: R & D Systems
MAB7788
102887-374EA 594.37 USD
102887-374
Anti-PTGIS Mouse Monoclonal Antibody [clone: 852307]
Antibodies
Simple Western

Type: Primary
Antigen: PTGIS
Clonality: Monoclonal
Clone: 852307
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR