Anti-CCDC134 Rat Monoclonal Antibody [clone: 815606]

Supplier: R & D Systems
MAB7786
102887-372EA 566.89 USD
102887-372
Anti-CCDC134 Rat Monoclonal Antibody [clone: 815606]
Antibodies
Type: Primary
Antigen: CCDC134
Clonality: Monoclonal
Clone: 815606
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR