Anti-TRIM28 Mouse Monoclonal Antibody [clone: 821912]

Supplier: R & D Systems
MAB7785
102887-370EA 594.37 USD
102887-370
Anti-TRIM28 Mouse Monoclonal Antibody [clone: 821912]
Antibodies
Simple Western

Type: Primary
Antigen: TRIM28
Clonality: Monoclonal
Clone: 821912
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR