Anti-CCDC134 Mouse Monoclonal Antibody [clone: 814208]

Supplier: R & D Systems
MAB7784
102887-368EA 566.89 USD
102887-368
Anti-CCDC134 Mouse Monoclonal Antibody [clone: 814208]
Antibodies
Type: Primary
Antigen: CCDC134
Clonality: Monoclonal
Clone: 814208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR