Anti-RFX6 Mouse Monoclonal Antibody [clone: 811704]

Supplier: R & D Systems
MAB7780
102887-366EA 577.02 USD
102887-366
Anti-RFX6 Mouse Monoclonal Antibody [clone: 811704]
Antibodies
Type: Primary
Antigen: RFX6
Clonality: Monoclonal
Clone: 811704
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR