Anti-NCAM2 Rat Monoclonal Antibody [clone: 125101]

Supplier: R & D Systems
MAB778
220021-543EA 763.57 USD
220021-543
Anti-NCAM2 Rat Monoclonal Antibody [clone: 125101]
Antibodies
Type: Primary
Antigen: NCAM2
Clonality: Monoclonal
Clone: 125101
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR