Anti-PFN1 Mouse Monoclonal Antibody [clone: 816536]

Supplier: R & D Systems
MAB7779
102887-364EA 577.02 USD
102887-364
Anti-PFN1 Mouse Monoclonal Antibody [clone: 816536]
Antibodies
Type: Primary
Antigen: PFN1
Clonality: Monoclonal
Clone: 816536
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR