Anti-TARDBP Mouse Monoclonal Antibody [clone: 671834]

Supplier: R & D Systems
MAB7778
102887-362EA 655.11 USD
102887-362
Anti-TARDBP Mouse Monoclonal Antibody [clone: 671834]
Antibodies
Type: Primary
Antigen: TARDBP
Clonality: Monoclonal
Clone: 671834
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR