Anti-ZG16B Mouse Monoclonal Antibody [clone: 817310]

Supplier: R & D Systems
MAB7777
102887-360EA 594.37 USD
102887-360
Anti-ZG16B Mouse Monoclonal Antibody [clone: 817310]
Antibodies
Type: Primary
Antigen: ZG16B
Clonality: Monoclonal
Clone: 817310
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR