Anti-FBXW7 Mouse Monoclonal Antibody [clone: 800201]

Supplier: R & D Systems
MAB7776
102887-358EA 633.42 USD
102887-358
Anti-FBXW7 Mouse Monoclonal Antibody [clone: 800201]
Antibodies
Type: Primary
Antigen Symbol: FBXW7
Antigen Name: F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase
Antigen Synonyms: AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
Clonality: Monoclonal
Clone: 800201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR