Anti-SIGLEC10 Rat Monoclonal Antibody [clone: 805903]

Supplier: R & D Systems
MAB7775
102887-356EA 594.37 USD
102887-356
Anti-SIGLEC10 Rat Monoclonal Antibody [clone: 805903]
Antibodies
Type: Primary
Antigen: SIGLEC10
Clonality: Monoclonal
Clone: 805903
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR