Anti-IL1F10 Rat Monoclonal Antibody [clone: 798036]

Supplier: R & D Systems
MAB7774
102887-354EA 594.37 USD
102887-354
Anti-IL1F10 Rat Monoclonal Antibody [clone: 798036]
Antibodies
Type: Primary
Antigen: IL1F10
Clonality: Monoclonal
Clone: 798036
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR