Anti-BAD Mouse Monoclonal Antibody [clone: 679431]

Supplier: R & D Systems
MAB7764
102887-352EA 577.02 USD
102887-352
Anti-BAD Mouse Monoclonal Antibody [clone: 679431]
Antibodies
Type: Primary
Antigen: BAD
Clonality: Monoclonal
Clone: 679431
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR