Anti-IGFBP6 Rat Monoclonal Antibody [clone: 122312]

Supplier: R & D Systems
MAB7761
220021-541EA 763.57 USD
220021-541
Anti-IGFBP6 Rat Monoclonal Antibody [clone: 122312]
Antibodies
Type: Primary
Antigen: IGFBP6
Clonality: Monoclonal
Clone: 122312
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR