Anti-IGFBP6 Rat Monoclonal Antibody [clone: 122316]

Supplier: R & D Systems
MAB776
220021-540EA 763.57 USD
220021-540
Anti-IGFBP6 Rat Monoclonal Antibody [clone: 122316]
Antibodies
Type: Primary
Antigen: IGFBP6
Clonality: Monoclonal
Clone: 122316
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR