Anti-SOX6 Mouse Monoclonal Antibody [clone: 667136]

Supplier: R & D Systems
MAB7759
102887-350EA 600.15 USD
102887-350
Anti-SOX6 Mouse Monoclonal Antibody [clone: 667136]
Antibodies
Type: Primary
Antigen: SOX6
Clonality: Monoclonal
Clone: 667136
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR