Anti-A1BG Mouse Monoclonal Antibody [clone: 875431]

Supplier: R & D Systems
MAB7757
102887-348EA 566.89 USD
102887-348
Anti-A1BG Mouse Monoclonal Antibody [clone: 875431]
Antibodies
Type: Primary
Antigen: A1BG
Clonality: Monoclonal
Clone: 875431
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR