Anti-IGFBP3 Rat Monoclonal Antibody [clone: 138202]

Supplier: R & D Systems
MAB775
220021-539EA 763.57 USD
220021-539
Anti-IGFBP3 Rat Monoclonal Antibody [clone: 138202]
Antibodies
Type: Primary
Antigen: IGFBP3
Clonality: Monoclonal
Clone: 138202
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR