Anti-ACVR1C Mouse Monoclonal Antibody [clone: 8156]

Supplier: R & D Systems
MAB77491
102887-346EA 594.37 USD
102887-346
Anti-ACVR1C Mouse Monoclonal Antibody [clone: 8156]
Antibodies
Type: Primary
Antigen: ACVR1C
Clonality: Monoclonal
Clone: 8156
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR