Anti-FSCN1 Mouse Monoclonal Antibody [clone: 833223]

Supplier: R & D Systems
MAB7745
102887-342EA 584.25 USD
102887-342
Anti-FSCN1 Mouse Monoclonal Antibody [clone: 833223]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: FSCN1
Antigen Name: fascin actin-bundling protein 1
Antigen Synonyms: FAN1, HSN, SNL, p55
Clonality: Monoclonal
Clone: 833223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR