Anti-ETV2 Rat Monoclonal Antibody [clone: 827940]

Supplier: R & D Systems
MAB7740
102887-340EA 577.02 USD
102887-340
Anti-ETV2 Rat Monoclonal Antibody [clone: 827940]
Antibodies
Type: Primary
Antigen: ETV2
Clonality: Monoclonal
Clone: 827940
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR