Anti-TNFSF8 Mouse Monoclonal Antibody [clone: 116615]

Supplier: R & D Systems
MAB774
220021-538EA 772.25 USD
220021-538
Anti-TNFSF8 Mouse Monoclonal Antibody [clone: 116615]
Antibodies
Type: Primary
Antigen: TNFSF8
Clonality: Monoclonal
Clone: 116615
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR