Anti-DBN1 Mouse Monoclonal Antibody [clone: 838102]

Supplier: R & D Systems
MAB7739
102887-338EA 561.11 USD
102887-338
Anti-DBN1 Mouse Monoclonal Antibody [clone: 838102]
Antibodies
Simple Western

Type: Primary
Antigen: DBN1
Clonality: Monoclonal
Clone: 838102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR