Anti-PD1 Rat Monoclonal Antibody [clone: 766104]

Supplier: R & D Systems
MAB7738
102887-336EA 633.42 USD
102887-336
Anti-PD1 Rat Monoclonal Antibody [clone: 766104]
Antibodies
Type: Primary
Antigen: PD1
Clonality: Monoclonal
Clone: 766104
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR