Anti-GJA1 Mouse Monoclonal Antibody [clone: 578618]

Supplier: R & D Systems
MAB7737
102887-334EA 594.37 USD
102887-334
Anti-GJA1 Mouse Monoclonal Antibody [clone: 578618]
Antibodies
Type: Primary
Antigen Symbol: GJA1
Antigen Name: gap junction protein, alpha 1, 43kDa
Antigen Synonyms: AVSD3, CMDR, CX43, EKVP, GJAL, HLHS1, HSS, ODDD, PPKCA
Clonality: Monoclonal
Clone: 578618
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR