Anti-IFNGR2 Hamster Monoclonal Antibody [clone: MOB47]

Supplier: R & D Systems
MAB773
220021-537EA 763.57 USD
220021-537
Anti-IFNGR2 Hamster Monoclonal Antibody [clone: MOB47]
Antibodies
Type: Primary
Antigen: IFNGR2
Clonality: Monoclonal
Clone: MOB47
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR