Anti-AMBP Mouse Monoclonal Antibody [clone: 784917]

Supplier: R & D Systems
MAB7724
102887-332EA 600.15 USD
102887-332
Anti-AMBP Mouse Monoclonal Antibody [clone: 784917]
Antibodies
Type: Primary
Antigen: AMBP
Clonality: Monoclonal
Clone: 784917
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR