Anti-BSG Rat Monoclonal Antibody [clone: 116318]

Supplier: R & D Systems
MAB772
220021-536EA 763.57 USD
220021-536
Anti-BSG Rat Monoclonal Antibody [clone: 116318]
Antibodies
Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: 116318
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR