Anti-MCAM Rat Monoclonal Antibody [clone: 733216]

Supplier: R & D Systems
MAB7718
102887-328EA 594.37 USD
102887-328
Anti-MCAM Rat Monoclonal Antibody [clone: 733216]
Antibodies
Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone: 733216
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR