Anti-IFI30 Mouse Monoclonal Antibody [clone: 844623]

Supplier: R & D Systems
MAB7715
102887-326EA 577.02 USD
102887-326
Anti-IFI30 Mouse Monoclonal Antibody [clone: 844623]
Antibodies
Type: Primary
Antigen: IFI30
Clonality: Monoclonal
Clone: 844623
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR