Anti-SIRT1 Mouse Monoclonal Antibody [clone: 834918]

Supplier: R & D Systems
MAB7714
102887-324EA 577.02 USD
102887-324
Anti-SIRT1 Mouse Monoclonal Antibody [clone: 834918]
Antibodies
Type: Primary
Antigen: SIRT1
Clonality: Monoclonal
Clone: 834918
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR