Anti-IL1R1 Hamster Monoclonal Antibody [clone: JAMA147]

Supplier: R & D Systems
MAB7711
220021-534EA 763.57 USD
220021-534
Anti-IL1R1 Hamster Monoclonal Antibody [clone: JAMA147]
Antibodies
Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: JAMA147
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR