Anti-IL1R1 Rat Monoclonal Antibody [clone: 129301]

Supplier: R & D Systems
MAB771
220021-533EA 772.25 USD
220021-533
Anti-IL1R1 Rat Monoclonal Antibody [clone: 129301]
Antibodies
Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: 129301
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR