Anti-SERPINA1A Rat Monoclonal Antibody [clone: 882213]

Supplier: R & D Systems
MAB7690
102887-314EA 594.37 USD
102887-314
Anti-SERPINA1A Rat Monoclonal Antibody [clone: 882213]
Antibodies
Type: Primary
Antigen: SERPINA1A
Clonality: Monoclonal
Clone: 882213
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR