Anti-PFKM Mouse Monoclonal Antibody [clone: 842735]

Supplier: R & D Systems
MAB7687
102887-312EA 594.37 USD
102887-312
Anti-PFKM Mouse Monoclonal Antibody [clone: 842735]
Antibodies
Simple Western

Type: Primary
Antigen: PFKM
Clonality: Monoclonal
Clone: 842735
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR