Anti-FLT3 Rat Monoclonal Antibody [clone: 113315]

Supplier: R & D Systems
MAB768
220021-530EA 763.57 USD
220021-530
Anti-FLT3 Rat Monoclonal Antibody [clone: 113315]
Antibodies
Type: Primary
Antigen Symbol: FLT3
Antigen Name: FMS-like tyrosine kinase 3
Antigen Synonyms: B230315G04, CD135, Flk-2, Flk2, Flt-3, Ly72, wmfl
Clonality: Monoclonal
Clone: 113315
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR