Anti-HDAC2 Mouse Monoclonal Antibody [clone: 806116]

Supplier: R & D Systems
MAB7679
102887-310EA 577.02 USD
102887-310
Anti-HDAC2 Mouse Monoclonal Antibody [clone: 806116]
Antibodies
Simple Western

Type: Primary
Antigen: HDAC2
Clonality: Monoclonal
Clone: 806116
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR