Anti-AKR1C3 Mouse Monoclonal Antibody [clone: 87171]

Supplier: R & D Systems
MAB7678
102887-308EA 600.15 USD
102887-308
Anti-AKR1C3 Mouse Monoclonal Antibody [clone: 87171]
Antibodies
Simple Western

Type: Primary
Antigen: AKR1C3
Clonality: Monoclonal
Clone: 87171
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR