Anti-CDH15 Rat Monoclonal Antibody [clone: 800512]

Supplier: R & D Systems
MAB76771
102887-306EA 577.02 USD
102887-306
Anti-CDH15 Rat Monoclonal Antibody [clone: 800512]
Antibodies
Type: Primary
Antigen: CDH15
Clonality: Monoclonal
Clone: 800512
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR