Anti-CDH15 Rat Monoclonal Antibody [clone: 800516]

Supplier: R & D Systems
MAB7677
102887-304EA 577.02 USD
102887-304
Anti-CDH15 Rat Monoclonal Antibody [clone: 800516]
Antibodies
Type: Primary
Antigen: CDH15
Clonality: Monoclonal
Clone: 800516
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR