Anti-FRMPD4 Mouse Monoclonal Antibody [clone: 809408]

Supplier: R & D Systems
MAB7676
102887-302EA 566.89 USD
102887-302
Anti-FRMPD4 Mouse Monoclonal Antibody [clone: 809408]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: FRMPD4
Antigen Name: FERM and PDZ domain containing 4
Antigen Synonyms: PDZD10, PDZK10
Clonality: Monoclonal
Clone: 809408
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR