Anti-BTRC Mouse Monoclonal Antibody [clone: 763524]

Supplier: R & D Systems
MAB7675
102887-300EA 577.02 USD
102887-300
Anti-BTRC Mouse Monoclonal Antibody [clone: 763524]
Antibodies
Simple Western

Type: Primary
Antigen: BTRC
Clonality: Monoclonal
Clone: 763524
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR