Anti-PDZD2 Mouse Monoclonal Antibody [clone: 787924]

Supplier: R & D Systems
MAB7670
102887-298EA 584.25 USD
102887-298
Anti-PDZD2 Mouse Monoclonal Antibody [clone: 787924]
Antibodies
Type: Primary
Antigen: PDZD2
Clonality: Monoclonal
Clone: 787924
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR